missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ VapLock™ Solvent Waste Manifold Caps for Bottles and Carboys

VapLock™ Solvent Waste Manifold Caps for Bottles and Carboys connect to waste containers for a secure closed system

481.00 € - 518.00 €

Specifications

For Use With (Application) Solvent Safety
For Use With (Equipment) Solvent-Compatible Containers
View More Specs

Products 2
Product Code Brand Material Size Thread Size (English) Price Quantity & Availability  
Product Code Brand Material Size Thread Size (English) Price Quantity & Availability  
15834594
View Documents
Cole-Parmer™
12018-84
PTFE DIN 45 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F)
518.00 €
Each
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
 
15844594
View Documents
Cole-Parmer™
12018-85
PTFE DIN 45 1/2 in. NPT(F), 1/4 in. NPT(F)
481.00 €
Each
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
 
Description

Description

  • Select from a variety of cap sizes to fit your container
  • Different materials, numbers and sizes of ports help customize to your application

Warranty

None

Specifications

Specifications

Solvent Safety
Solvent-Compatible Containers
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.