missing translation for 'onlineSavingsMsg'
Learn More

Watson-Marlow 323 Series Drive with 313DW Pumphead

300 series cased pumps. Single or multi-channel benchtop pumps with manual, remote, analogue, RS232 control and accurate dispensing.

Brand:  Watson-Marlow 030.3144.3DU

10045.19 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 12426350

  • 11750.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • 300 series laboratory pumps are a benchtop pumping solution with manual, remote, analogue or RS232 control capabilities. Featuring a chemically resistant ABS top and IP31 protection, 323E/D pumps have a membrane keypad that controls the pump, and a digital LCD to display speed. There are four models with increasing degrees of functionality:
  • 323E/DDu RS232/auto/manual enhanced-control high-flow pumps
  • 323E/DU and 323E/DU analogue-auto and enhanced manual-control pumps
  • 323E/DS high-performance manual control pumps
  • 323E/DE manual control pumps

Pumphead options available as standard (others available on request):

  • 313D and 313D2 three-roller pumpheads for continuous tubing
  • 314D and 314D2 four-roller pumpheads for continuous tubing
  • 314MC four-roller five-channel microcassette pumpheads
  • 318MC eight-roller five-channel microcassette pumpheads
  • 501RL and 501RL2 pumpheads for continuous tubing
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Analog
IP31
3 to 400 rpm
Run/Stop, direction change via contact closure or 5V TTL; Remote operation of MemoDose (hand/foot switch)
<70 dB (A) at 1m
313DW
104°F
40°C
230 x 230 x 132 mm
100/120/220/240 V
LCD
0.81 to 2000 mL/min.
Yes
323U/D
3
3 to 400rpm 133:1; 1.5 to 220rpm 147:1
40°F
4°C
100 VA
50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.