Learn More
Macherey-Nagel™ VarioPrep™ Guard Column Holder for Preperative HPLC
Use VarioPrep™ stainless steel columns for HPLC. VarioPrep™ Guard Column Holder for Preperative HPLC come in different sizes for a variety of columns.
Brand:  Macherey-Nagel™ 718253
 View more versions of this product
Product Code. 12757690
  • 500.00 € / Each
Estimated Shipment: 12-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Features
  • Column system for preparative HPLC manufactured from stainless steel with two adjustable end fittings, e.g., for frequent use of backflushing techniques
  • Allows compensation of a dead volume, which could result at the column inlet after some time of operation, without need for opening the column
  • Can be packed with all NUCLEODUR™ and NUCLEOSIL™ spherical silicas
  • Column holders come in a variety of sizes for different columns
Specifications
Specifications
32 mm
Guard Column Holder
15 mm