Learn More
Macherey-Nagel™ VarioPrep™ Nucleodur™ C18 Isis HPLC Columns
Brand:  Macherey-Nagel™ 762405.100
 View more versions of this product
Product Code. 12449946
  • 1330.00 € / Each
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
Specifications
Preparative Column
Steroids, (o,p,m-) Substituted Aromatics, Fat-Soluble Vitamins
5800 psi (400 bar)
1 to 10
340 m²/g
L1
Silica
Steric Selectivity
Reversed Phase
10 mm
125 mm
5 μm
110 Å
60°C
C18 Isis
20%
Yes
VarioPrep HPLC Column