missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Varioskan™ ALF Multimode Microplate Reader

The Thermo Scientific Varioskan ALF Multimode Microplate Reader is a versatile, entry level reader for most common absorbance, turbidimetric, fluorescence, and luminescence microplate assays.

Brand:  Thermo Scientific™ VA000010C

 View more versions of this product

Product Code. 30178790

  • 23690.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Thermo Scientific Varioskan ALF Multimode Microplate Reader is a versatile, entry level reader for most common absorbance, turbidimetric, fluorescence, and luminescence microplate assays. Its photometric optical design is based on a xenon flash lamp and monochromator, enabling continuous wavelength selection from UV through visible range up to 1000 nm. The broad wavelength range enables many common assays, including nucleic acid and protein quantification, bacterial growth curves, ELISA, and cell viability.

Enhance your fluorescence and luminescence measurements with the filter-based optics

With the Varioskan ALF reader, fluorescence and luminescence measurements are performed with filter-based optics for optimal specificity and sensitivity. The system comes with three pre-installed excitation and emission filter sets suitable for many common fluorometric applications like Quant-IT DNA quantitation assays and PrestoBlue and AlamarBlue cell viability assays. Additionally, five open filter positions are available for customized fluorometric and luminometric measurements. The filter slots are easy to access on the instrument's side with no tools required for filter exchange. Also, commonly used FRET and BRET assays are easy to set up and perform thanks to Varioskan ALF reader's unique filter-based optics design.

This package includes a Varioskan ALF Multimode Microplate Reader (Cat. No. VA000010C) and a Rapid Exchange two-year extended warranty.

Achieve consistently accurate results with automatic gain adjustment

The automatic gain adjustment feature selects the ideal fluorometric or luminometric reading range for your instrument based on signal intensity in the well, eliminating the need to manually adjust measurement parameters. The results are consistent no matter what signals are measured.

Additional features

  • Kinetic and endpoint measurements for a variety of applications
  • Spectral scanning for photometric measurements
  • Separate optimized measurement modes for absorbance and turbidimetric measurements
  • A self-diagnostics system that includes a set of initialization tests and adjustments, ensuring instrument readiness for operation
  • Compatible with 6- to 384-well microplates with and without lids
  • Reads μDrop and μDrop Duo plates for microvolume analysis of DNA, RNA, and proteins
  • Performs onboard shaking and incubation for temperature-critical assays
  • Operated through SkanIt software
    • Unlimited installation on multiple computers, no annual fee required
    • Includes an extensive cloud-based library of ready-made protocols
    • Two editions available: Research Edition for life sciences and Drug Discovery Edition with features for compliance with 21 CFR Part 11
    • Multiple user interface languages available
    • Automation compatibility for integration with automation interface
  • Thermo Scientific verification plates provide comprehensive and traceable validation of the instrument optical performance. Ready-made SkanIt software protocols provide easy to read reports with defined measurement parameter limits

Varioskan ALF Multimode Microplate Reader with 2-year Extended Warranty (A66711), REX is a Varioskan ALF specific package SKU including the instrument (VA000010C), and an extended warranty to 2 years which includes a rapid exchange service (REX) should the instrument get broken during the warranty period.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Multimode Microplate Reader
Photomultiplier Tube (PMT)
Ambient +2° to 45°C
Xenon Flash Lamp
Varioskan
Linear, Orbital, Double Orbital
≤16 sec. (Fast mode)
±1.0% + 0.003 Abs (0 to 2 Abs), ±2.0% (2.0 to 2.5 Abs) at 450 nm (96-well plate)
61 cm
210-720 nm
Three factory installed fluorometric filter pairs: Coumarin family (Excitation 345/25 nm, Emission 450/40 nm); Fluorescein family (Excitation 485/15 nm, Emission 525/15 nm); Resorufin family (Excitation 555/15 nm, Emission 615/45 nm)
40 cm
0 to 2.5 Abs, 2.0% at 450 nm (96-well plate)
Photometry (DNA): <1 ng/μL (96-well half area plate); Turbidimetry (formazin): <5 turbidity units (FTUs) (96-well plate); Fluorometry (fluorescein): 0.35 fmol/well (black 384-well plate); Luminometry (ATP): <275 amol/well (white 96-well plate)
100/115/220/230 V
24.5 kg
1 each
No
61 x 34 x 40 cm
PC Software (Thermo Scientific SkanIt Software)
Photometry: ≤10 sec. (fast mode); Fluorometry and Luminometry: ≤20 sec. (96-well plate)
Endpoint, Kinetic
SkanIt Software
200 to 1000 nm
Photometry: 2.5 nm
Fluorometry and Luminometry: ≥6 Decades
200-710 nm
Photometric, fluorometric and luminometric microplate assays
SkanIt software, Research Edition
SD <0.003 Abs, or CV <1.0% at 450 nm
English, French, German, Spanish, Portuguese, Italian, Japanese, Chinese
Standard
34 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.