Learn More
Eppendorf™ Varispenser™ 2x Bottle-Top Dispenser with Recirculation Valve
Ideal for dispensing aliquots of liquid from large supply bottles.
389.00 € - 708.00 €
Specifications
Channels 1
View More Specs
Products 6
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
15787367
View Documents
Eppendorf™
4967000014
0.2 to 2.0 mL
389.00 €
Each
In Stock
to see stock.
Add to basket
15797367
View Documents
Eppendorf™
4967000022
0.5 to 5.0 mL
389.00 €
Each
In Stock
to see stock.
Add to basket
15707377
View Documents
Eppendorf™
4967000030
1.0 to 10 mL
389.00 €
Each
In Stock
to see stock.
Add to basket
15717377
View Documents
Eppendorf™
4967000049
2.5 to 25 mL
523.00 €
Each
In Stock
to see stock.
Add to basket
15781587
View Documents
Eppendorf™
4967000057
5.0 to 50 mL
563.00 €
Each
In Stock
to see stock.
Add to basket
15727377
View Documents
Eppendorf™
4967000065
10 to 100 mL
708.00 €
Each
In Stock
to see stock.
Add to basket
Description
Description
Ideal for dispensing aliquots of liquid from large supply bottles.
  • Recirculation valve
  • Standard GL 45 thread on all sizes for most common bottle threads
  • Safety valve balls in discharge valve prevents leakage when discharge tube is not mounted
  • Screwable air vent cap for easy assembly of drying tube or microfilter
  • Oval-shaped dispenser housing for easy transportation
  • PFA seal of the sliding piston prevents seizing up
  • Fixation slide on interior toothed track for rapid and accurate volume adjustment
  • High chemical resistance of all dispensing components
  • Telescopic aspirating tubes for variable adjustment to bottle height
  • Fully autoclavable without disassembly
  • User adjustment option
Specifications
Specifications
1