missing translation for 'onlineSavingsMsg'
Learn More

Medline™ Varizoom 6 Stereomicroscope with LED Illumination

Use this microscope for a variety of applications. Medline™ Varizoom 6 Stereomicroscope with LED Illumination features a powerful processing function for real-time preview, multiple peripheral interfaces and dynamic calibrating and measuring.

Brand:  Medline™ 7700.0006

Product Code. 12683025

  • 3525.00 € / Each
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Robust Stereo Zoom microscope with 1:8 zoom ratio and double LED illumination
  • Choice of 3 zoom ratios 1:6, 1:8 or 1:10
  • Magnification range 2x to 320x
  • Planachromatic objectives
  • Integral one piece column stand to eliminate vibration and image distortion
  • LED cold light variable intensity transmitted and incident light sources
  • Binocular configuration, with options for a beam splitter arm to facilitate digital recording when required
Specifications

Specifications

220/240 V
22 mm
220/240 V 50/60 Hz
Binocular, 20° Inclined
10X
8X to 50X, 8X to 64X, 8X to 80X
1:6
Stereoscopic Zoom Microscope
50/60 Hz
1 Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.