missing translation for 'onlineSavingsMsg'
Learn More

Medline Scientific™ Varizoom LED Binocular Zoom Stereomicroscope

Product Code. 12693025
Click to view available options
Magnification Power:
8X to 64X
8X to 80X
Zoom Range:
0.047222222222
0.048611111111
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12693025

Brand: Medline Scientific™ 7700.0008

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

A stereoscopic zoom microscope with parallel infinity optics and an optical zoom system.

  • Binocular configuration as standard
  • Three zoom ranges are available
  • Binocular optical head inclined at 20°
  • Interpupillary distance adjustment (Siedentopf type) 45 to 75mm
  • Dioptric compensation on both eyepiece tubes
  • Includes 2 rubber eyecups, one blue filter, dust cover and instruction manual

Specifications

Includes 2 rubber eyecups, one blue filter, dust cover and instruction manual
Voltage 220/240 V
Magnification Power 8X to 64X
Objective Types Plan Achromatic
Zoom Range 0.047222222222
Electrical Requirements 220/240 V 50/60 Hz
Product Type Stereoscopic Zoom Microscope
Head Style Binocular, 20° Inclined
Lighting Type LED
Frequency 50/60 Hz
Eyepiece WF 10X/22 mm, Pointer in One Eyepiece
Quantity 1 Each
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.