Learn More
Sonics Materials VC-505 Ultrasonic Processor
The VC-505 Ultrasonic Processor is extremely capable and versatile, it can safely process a wide range of organic and inorganic materials – from microliters to liters.
Brand:  Sonics Materials VC-505-220
Product Code. 10532103
  • 5345.60 € / Each
Estimated Shipment: 08-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Extremely capable and versatile, it can safely process a wide range of organic and inorganic materials - from microliters to liters
  • 500 and 750 Watt Ultrasonic Processors to safely process a wide range of organic and inorganic materials, from 250 microliters to 1 liter
  • Typical applications include nanotechnology (producing nanoparticle materials and Graphene dispersions), cell lysing, sample preparation, homogenization, ChIP Assay, emulsification, disaggregation and deagglomeration, plus uses in the field of sonochemical liquid processing
  • Energy monitor, digital wattmeter, microprocessor-based and programmable
  • 10 hour process timer
Specifications
Specifications
Processing a wide range of organic and inorganic materials, from 250 uL to 1 L
0.5 in. (13 mm) probe with replaceable tip, tool kit, and instruction manual
10 hr. process timer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Sonics Materials VC-505 Ultrasonic Processor > VC-505 Ultrasonic Processor; Wattage: 505 W