Learn More
Sonics Materials VCX 500 Ultrasonic Microprocessor
Safely processes a wide range of organic and inorganic materials, from 250 microliters to 1 liter.
Brand:  Sonics Materials VCX-500-220
Product Code. 11753619
  • 4715.00 € / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ultrasonic processors for small and medium volume applications
  • Safely processes a wide range of organic and inorganic materials, from 250 microliters to 1 liter 
  • Typical applications include nanotechnology (producing nanoparticle materials and Graphene dispersions), cell lysing, sample preparation, homogenization, ChIP Assay, emulsification, disaggregation and deagglomeration, plus uses in the field of sonochemical processing
  • Energy and temperature set-point control, digital wattmeter, automatic tuning, and 10 hour process timer
Specifications
Specifications
500 W
Nanotechnology (producing nanoparticle materials and Graphene dispersions), cell lysing, sample preparation, homogenization, ChIP Assay, emulsification, disaggregation and deagglomeration, plus uses in the field of sonochemical processing
10 hour process timer
13.5 x 7.5 x 9.25 in. (340 x 190 x 235 mm)
Standard 0.5 in. diameter probe with replaceable tip
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products