Learn More
Sonics Materials VCX 750 Ultrasonic Microprocessor
The 750 Watt Ultrasonic Processor is for small and medium volume applications and is ideal for safely processing a wide range of organic and inorganic materials from 250 microliters to 1 liter.
Brand:  Sonics Materials VCX-750-220
Product Code. 11763619
  • 5875.00 € / Each
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ultrasonic processors for small and medium volume applications
  • 500 and 750 watt ultrasonic processors to safely process a wide range of organic and inorganic materials, from 250 microliters to 1 liter
  • Typical applications include nanotechnology (producing nanoparticle materials and Graphene dispersions), cell lysing, sample preparation, homogenization, ChIP Assay, emulsification, disaggregation and deagglomeration, plus uses in the field of sonochemical liquid processing
  • Energy monitor, digital wattmeter, microprocessor-based and programmable
  • 10 hour process timer
Specifications
Specifications
750 W
Small and medium volume applications
10 hr. process timer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products