missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Venturi Vacuum Pump

The remarkably compact size of these Cole-Parmer Venturi Vacuum Pump enables you to design vacuum systems with smaller, less expensive components and reduced maintenance costs. With no moving parts or electrical hazards, these air-powered pumps are also intrinsically safe.

Brand:  Cole-Parmer™ 78165-20

Product Code. 11720169

  • 353.00 € / Each
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • No moving parts ensures long life with low maintenance.
  • Venturi pumps are air-powered, require no electrical power source or lubricant.
  • Place these compact vacuum pumps close to application for fast response and minimal vacuum loss.
Specifications

Specifications

65.1 Mbar
Venturi Vacuum Pump
140°F
90.6 L/min.
11.75 x 2.54 x 8.89 cm (4.625 x 1 x 3.5 in.)
Aluminum, Brass, Buna N
60°C
Vacuum System
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.