Learn More
Veolia Water Systems™ UV Lamp
Rich, robust UV lamp that features UV light acting as a bactericide to break down and photo-oxidize organic contaminants to polar or ionized species.
Brand:  Veolia Water Systems™ UVEGBU2009
 View more versions of this product
Product Code. 13175950
  • 231.00 € / Each
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • UV light is used as a bactericide and to break down and photo-oxidise organic contaminants to polar or ionized species
  • The UV bactericidal action damages DNA and RNA polymerase at low doses thereby preventing replication
Specifications
Specifications
UHQ6 Ultrapure Water System