missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals (±)-Verapamil hydrochloride, 99+%

Product Code. 10441012
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152-11-4
Molecular Formula C27H38N2O4·HCl
Molecular Weight (g/mol) 491.07
MDL Number MFCD00055208
InChI Key DOQPXTMNIUCOSY-UHFFFAOYSA-N
Synonym verapamil hydrochloride, verapamil hcl, manidon, calcan hydrochloride, cardibeltin, cardiabeltin, cardioprotect, veroptinstada, calaptin, caveril
PubChem CID 62969
IUPAC Name 2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]-2-propan-2-ylpentanenitrile;hydrochloride
SMILES CC(C)C(CCCN(C)CCC1=CC(=C(C=C1)OC)OC)(C#N)C2=CC(=C(C=C2)OC)OC.Cl

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 140.0°C to 144.0°C
Color White
pH 4.5 to 6.5 (5% soln.)
Ignition Residue 0.1% max.
Loss on Drying 0.5% max. (105°C, 2 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 10144
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in ethanol
Formula Weight 491.07
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material (±)-Verapamil hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • (±)-Verapamil hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.