missing translation for 'onlineSavingsMsg'
Learn More

Veratrole, 99+%, Thermo Scientific Chemicals

Product Code. 10225590
100 g, Glass bottle
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-16-7
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00008357
InChI Key ABDKAPXRBAPSQN-UHFFFAOYSA-N
Synonym veratrole, veratrol, pyrocatechol dimethyl ether, o-dimethoxybenzene, catechol dimethyl ether, benzene, 1,2-dimethoxy, 2-methoxyanisole, o,o-dimethyl catechol, 2-dimethoxybenzol, benzene, o-dimethoxy
PubChem CID 7043
ChEBI CHEBI:59114
IUPAC Name 1,2-dimethoxybenzene
SMILES COC1=CC=CC=C1OC

Specifications

Melting Point 15°C
Color White to Cream
Density 1.0800g/mL
Boiling Point 206°C to 207°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C6H4(OCH3)2
Refractive Index 1.5330 to 1.535
Quantity 100 g
Beilstein 06, 771
Merck Index 15, 10150
Specific Gravity 1.08
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol,methanol,acetone and ether
Formula Weight 138.17
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material Veratrole, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Veratrole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Veratrole, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.