Learn More
Linden™ Vernier Callipers with Micrometer Screw Gauge
Product Code. 13025903
Click to view available options
Length (Metric):
115 mm
150 mm
Unit Size:
Each
Material:
Plastic
Product Code. 13025903
Brand: Linden™ H.DR39500
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Can be used for internal, external and depth measurement. Linden™ Vernier Callipers with Micrometer Screw Gauges have a micrometer screw gauge supplied in case.
  • General purpose with thumb wheel for fine adjustment
  • Metric and imperial dual graduations
  • Available in a range of lengths
Specifications
Length (Metric) 150 mm