Learn More
R&D Systems™ VersaClone cDNA, Human GlyT2/SLC6A5 (NP_004202)
SLC6A5 is found in glycinergic axons and maintains a high presynaptic pool of neurotransmitter at glycinergic synapses
Brand: R&D Systems™ RDC1231
This item is not returnable.
View return policy
Description
The Human GlyT2/SLC6A5 (NP_004202) VersaClone cDNA clone contains the complete ORF for GlyT2/SLC6A5, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The GlyT2/SLC6A5 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature
Specifications
NP_004202 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
10 μg at 0.2 μg/μL | |
2406bp | |
Human |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Reasearch Use Only; Not for Use in Humans
Spot an opportunity for improvement?Share a Content Correction