missing translation for 'onlineSavingsMsg'
Learn More

3M™ Versaflo™ Air Cooling Regulator, V-100E

3M™ Versaflo™ Air Cooling Regulator V-100E can cool supplied air down to 28°C while providing protection against particulates, gases and vapours, or combination hazards.

Brand:  3M™ 7000103980

577.83 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


Product Code. 15306057

  • 740.00 € / Each
Estimated Shipment: 06-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Individually cool your incoming supplied air down to 28°C
  • Ideal for workers exposed to warm temperatures in addition to requiring protection against particulates, gases and vapours or combination hazards
  • Integrated warning whistle indicates low air flow
  • Adjustable temperature controller
  • Working pressure of 4 to 6 bar
  • Air volume of 170 to 600 L/min
  • Includes a belt-mounted insulating pad
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CE Approved
Supplied Air Regulator
0.346 m
0.086 m
Black
V-100E
0.118 m
650 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers