Learn More
3M™ Versaflo™ Battery Attachment Tool TR-838/94244(AAD)
3M™ Versaflo™ Battery Attachment Tool TR-838/94244(AAD) is an accessory for attaching TR-830 batteries to the TR-802E Powered Air Turbo.
Brand: 3M™ 7100150921
16.51 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Enables attachment of TR-830 battery to TR-802E Powered Air Turbo
- Ensures secure fastening
- Prevents accidental battery removal and spark risk during use
- The mechanical fastening action prevents accidental battery removal and reduces spark risk during use per EN 60079-11
- Intrinsically safe and suitable for potentially explosive atmospheres
Specifications
CE Approved, NIOSH Approved | |
Battery Attachment Tool | |
TR-802E Powered Air Turbo |