Learn More
3M™ Versaflo™ Breathing Tubes for Headtops and Air Delivery Units
Lightweight BT-series breathing tubes are compatible with all 3M™ Versaflo™ headtops and air delivery units.
57.09 € - 67.00 €
Specifications
Product Type Breathing Tube
For Use With (Application) Assembly, facility maintenance, grinding, machine operation, painting, sanding
For Use With (Equipment) Versaflo™ PAPR Systems
View More Specs
Products 2
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
10099369
View Documents
3M™
7000000158
Small/Medium
67.00 €
Each
Estimated Shipment: 18-02-2025
to see stock.
Add to basket
10566490
View Documents
3M™
7000000159
Medium/Large This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Long, lightweight breathing tube that is easy to wipe down
  • Quick release swivel enables one-handed connection and release
  • Disposable breathing tube cover is available to help cleaning and decontamination operations
  • 3M Catalog# BT-20S and BT-20L
Specifications
Specifications
Breathing Tube
Assembly, facility maintenance, grinding, machine operation, painting, sanding
Versaflo™ PAPR Systems