missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Versaflo™ Comfort Pad for S-950 Premium Head Suspension
For use with S-950 Premium Head Suspension
16.25 € - 28.65 €
Specifications
Color | Gray |
---|---|
Disposable | Reusable |
Product Type | Head Suspension |
For Use With (Application) | Cleaning, Electrical, Facility Maintenance, Welding |
Description
- For use with S-950 Premium Head Suspension
- Wrapped around headband to create more comfortable fit and to absorb sweat
- 3M Catalog# S-953 , 3M Catalog# S-954
Specifications
Gray | |
Reusable | |
Head Suspension | |
Cleaning, Electrical, Facility Maintenance, Welding |