Learn More
3M™ Versaflo™ Easy Clean Backpack
Secure your 3M™ Powered Air Respirator on your back with 3M™ Powered Air Respirator Back Pack Harnesses.
Brand: 3M™ 7100300071
127.08 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Description
- Hold your 3M™ Powered Air Turbo firmly in place with 3M™ Powered Air Respirator Back Pack Harnesses.
- Our backpack harnesses is specifically designed to provide a secure hold while you work, enabling the turbo unit to be worn like a backpack rather than waist mounted.
Specifications
Black | |
Various | |
Standard |
Reusable | |
Backpack |