Learn More
3M™ Versaflo™ M-Series Faceshields and Helmets
Product Code. 17825666
Click to view available options
Includes:
Comfort Faceseal
Flame Resistant Faceseal
Flame Resistant Poly Faceseal
Flame Resistant Shroud
Highly Durable Shroud
Unit Size:
Each
Model:
M-406
Product Code. 17825666
Brand: 3M™ 7100258667
3M™ Versaflo™ M-Series Faceshields and Helmets can help protect your workers from a variety of respiratory and impact hazards while maintaining comfort. They are are lightweight, compact and easy to maintain.
  • M-Series are specifically designed to work together with your 3M™ Versaflo™ Powered Air Respirator and 3M™ Supplied Air Respirator Systems.
  • Deflector allows users to direct the airflow inside the headtop to increase comfort.
  • Fully-adjustable suspension for customized fit and weight distribution.
  • Accessory options availalble for protection to help handle the environments you encounter.
  • Visor combines good peripheral and downward vision.
  • Specific headtops can be used with a range of 3M™ Peltor™ hearing protection as well as an attachable microphone for ease of communication.
Specifications
Color Light Gray
Design Type Comfort Faceseal
Disposable Reusable
Filter Type None
Material Polyethylene
Product Type Hood with Faceseal
Size Standard
With Exhalation Valve No
Includes Comfort Faceseal