missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Versaflo™ M-Series Faceshields and Helmets
3M™ Versaflo™ M-Series Faceshields and Helmets can help protect your workers from a variety of respiratory and impact hazards while maintaining comfort. They are are lightweight, compact and easy to maintain.
Brand: 3M™ 7100257937
432.90 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
View more versions of this product
Description
- M-Series are specifically designed to work together with your 3M™ Versaflo™ Powered Air Respirator and 3M™ Supplied Air Respirator Systems.
- Deflector allows users to direct the airflow inside the headtop to increase comfort.
- Fully-adjustable suspension for customized fit and weight distribution.
- Accessory options availalble for protection to help handle the environments you encounter.
- Visor combines good peripheral and downward vision.
- Specific headtops can be used with a range of 3M™ Peltor™ hearing protection as well as an attachable microphone for ease of communication.
Specifications
Black | |
Reusable | |
Polyurethane-coated Polyamide | |
Standard | |
Comfort Faceseal |
Comfort Fit | |
None | |
Respirator Helmet | |
No | |
Polycarbonate Visor |