Learn More
3M™ Versaflo™ Over-visor, M-967
Over-visor to protect against UV/IR, metal sparks and splashes.
Brand:  3M™ 7100099909
Product Code. 17277640
  • 278.00 € / Each
Estimated Shipment: 06-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Gold IR/UV over-visor
  • Protects against UV/IR, and metal sparks and splashes
  • 500 x 250 mm
  • For use with 3M™ Versaflo™ Over-visor Frame, M-961
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Gold
3 M™ Versaflo™ Over Visor Frame M 961 N
0.503 m
0.012 m
Headgear Cover
1/Cs.
0.252 m