Learn More
3M™ Versaflo™ PAPR Breathing Tube Covers
PAPR disposable breathing covers are designed to help keep Versaflo™ PAPRs clean from debris.
Brand: 3M™ 7100260719
This item is not returnable.
View return policy
Description
- Made from a liquid resistant material with zippers for convenient donning and doffing
- Clear plastic window for control viewing and button activation
- Helps keep PAPR clean from debris
- Compatible with Versaflo™ PAPRs
- Popular for paint spray application
- Made with no natural rubber latex components
Spécification
Plastic | |
Protects PAPR blower from dirt and debris | |
TR-381 |
Breathing Tube Cover | |
TR-300+ PAPR | |
10/Cs. |