Learn More
3M™ Versaflo™ Powered Air Respirator System Healthcare Ready Kit, TR-300E+ HKL
3M™ Versaflo™ TR-300+ Powered Air Respirators are versatile and easy-to-use respiratory protection against particulates hazards.
Brand: 3M™ 7100233765
1079.49 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Slim, lightweight unit is well-balanced and comfortable to wear for long work shifts
- Two user selectable airflows enable the user to control flow settings to allow the user to find the setting they find most comfortable
- Powered Air Respirators automatically adjust airflow to compensate for filter loading to deliver consistent filtered air during use
- Audible and visual alerts for low airflow and low battery charge-level to keep you prepared and informed
- Able to be worn on a belt or backpack
- Available in convienient, easy to buy assemblies and kits
Specifications
9 to 12 hr. | |
PARTICULATE | |
PAPR Healthcare Systems and Kits | |
TR-3712E Filter | |
No |
High Capacity Lithium Ion - Rechargeable | |
Black | |
One Size | |
S-133L Headcover |