missing translation for 'onlineSavingsMsg'
Learn More

3M™ Versaflo™ S-Series Headcovers and Hoods

3M™ Versaflo™ S-Series Headcovers and Hoods offer a variety of options for faceseals, collars, shrouds, and suspensions, allowing you to choose the respiratory protection that provides the most value and comfort for your team.

Brand:  3M™ 7100079870

 View more versions of this product

Product Code. 11711143

  • 181.00 € / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • S-Series respiratory headcovers and hoods are specifically designed to work together with your Versaflo™ Powered Air Respirator and 3M™ Supplied Air Respirator Systems.
  • Provides head, face, neck and shoulder coverage from dust and paint spray.
  • Provides a good field of view.
  • Helps protect eyes and face from liquid splash and low energy flying particles and dust.
  • Versatile accessories available that can help you adapt the hood to your particular environment.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1000
Premium Replacement Hood with Inner Collar, S-655
Polypropylene
Assembly, facility maintenance, machine operation, painting, sanding, pharmaceutical manufacturing, liquid handling, battery manufacturing and recycling, petrochemical refining and extraction
S-605 Hood with Inner Collar, S-950 Premium Head Suspension, S-955 Airflow Adjustment Cover, S-954 Size Reducing Pad, S-922 Peel-Off
S-655
Premium Resusable Suspension
15.5 in.
10.19 in.
8.88 in.
NIOSH
White
Hood with Neck Collar
3M™ powered air purifying respirator and supplied air respirator valves
PETG
1 Ea.
15.5 x 10.19 x 8.88 in. (39.37 x 25.88 x 22.55 cm)
39.37 cm
25.88 cm
22.55 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.