Learn More
3M™ Versaflo™ Powered Air Respirator Starter Kit, TR-619E
3M™ Versaflo™ Powered Air Turbo Starter Kit, TR-619E is our ready-to-use kit for respiratory protection against particles, gases and vapours. It comes with everything you need to get started. Simply add your 3M™ Versaflo™ Headtop for complete respiratory protection.
Brand: 3M™ 7100053532
1552.52 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Simple starter kit with all the component parts to get you up and running. Simply add the headtop required
- Easy to use, versatile respiratory protection against dusts, mists, fumes and organic vapours
- Kit includes Turbo Unit TR-602E, A2P filter, filter cover, pre-filters, spark arrestors, easy clean belt, high capacity battery, single station battery charger kit, BT-30 length adjusting breathing tube and airflow indicator
- Compatible headtop required
- Visual, vibratory and audible warning alarms to alert to low battery and air flow
- System run time of 9.5-19 hours depending on battery, flow rate, headtop and filter
Specifications
PAPR Assembly, No Headtop | |
Versaflo™ |
Videos
-
3M Versaflo powered air turbo TR-600
Product Content Correction
Product Title
3M™ Versaflo™ Powered Air Respirator Starter Kit, TR-619E > Product Type: PAPR System Kit, Color: Light Gray, Model: TR-619E
Spot an opportunity for improvement?Share a Content Correction