Learn More
Ansell™ VersaTouch™ 37-501 Series Blue Nitrile Immersion Gloves Product Code.: 15324092
Add to basket
Ansell™ VersaTouch™ 37-501 Series Blue Nitrile Immersion Gloves
Product Code. 15324092 Shop All Ansell Products
10.5
Click to view available options
Size:
6.5
7.5
8.5
9.5
10.5
Product Code. 15324092
Brand: Ansell™ 37501105
Edge
Add to basket
Add to basket
Resists many solvents and oil based chemicals. Ansell™ VersaTouch™ 37-501 Series Blue Nitrile Immersion Gloves have good abrasion resistance.
  • Excellent resistance to many solvents and oil based chemicals
  • Very good abrasion resistance
  • Flexible and comfortable for an excellent dexterity
  • Gauntlet cuff style
  • Raised diamond grip
  • Powder free
TRUSTED_SUSTAINABILITY
Specifications
Back Material Nitrile
Cuff Material Nitrile
Product Type Immersion Gloves
Size 10.5
Style 37-501
Thickness (Metric) 0.42 mm
Glove Cuff Color Blue
Glove Cuff Style Gauntlet
Liner Material Cotton Flocking
Quantity 12 Pairs
Resistant To Excellent resistance to many solvents and oil based chemicals
Series 37-501
Product Title
Ansell™ VersaTouch™ 37-501 Series Blue Nitrile Immersion Gloves >