missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ VersaTouch™ 92-205 Series Single-Use White Nitrile Gloves

Product Code. 12697051
Click to view available options
Size:
7
8
9
10
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 12697051

Brand: Ansell™ 92205100

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Improved manufacturing processes for longer wear, greater tear strength and better grip in dry, wet and fatty conditions. A thinner glove with enhanced fingertip flexibility and improved overall comfort.

  • Single use gloves
  • Improved manufacturing processes for longer wear, greater tear strength and better grip in dry, wet and fatty conditions
  • Thinner glove with enhanced fingertip flexibility and improved overall comfort
  • Textured fingertips provide good comfort and tactility for sensitive, precise handling
  • Latex-free: no risk of Type 1 allergies
  • Approved for food contact applications
  • Ideal for handling all fatty food

Applications:

  • Prepared meals
  • Fruit and vegetable processing
  • Dairy processing
  • Food distribution
  • Catering
TRUSTED_SUSTAINABILITY

Specifications

Accelerator Free No
Applications Industrial
AQL 1.5
Color White
Disposable Single-use
Exterior Finish Fully Textured
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Nitrile Gloves
Size 10
Sterility Non-sterile
Thickness (English) 2.9 mil
Thickness (Metric) 0.07 mm
Model 9220510
Series 92-205
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.