missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Versene Solution
GREENER_CHOICE

Versene Solution

Brand:  Gibco™ 15040033

Product Code. 11518876

  • 20.49 € / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Versene is an EDTA solution for use as a gentle non-enzymatic cell dissociation reagent. Gibco™ Versene Solution (0.48 mM) is formulated as 0.2 g EDTA(Na4) per liter of Phosphate Buffered Saline (PBS).

Product Use
For Research Use Only. Not for use in diagnostic procedures.

Dual-site cGMP Manufacturing and Quality System
Gibco™ Versene Solution is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offers an identical Gibco™ Versene Solution product made in our Grand Island facility (15040-066). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ISO 13485 standards, FDA
Versene Solution, 100mL, Liquid, 1:5000 Dilution, EDTA Chelators, No Phenol Red, Contains 0.2 g EDTA(Na4) per Liter of Phosphate Buffered Saline (PBS), Water Soluble, FDA and ISO 13485 Standards, Used for Mammalian Cell Culture, 18 Months Shelf Life
Liquid
EDTA
Mammalian cell culture
For research use only
2 to 8°C
Mammalian Cells
Cell Culture Dissociation Reagent
No Phenol Red
100 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Versene Solution > 100mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.