Learn More
Thermo Scientific™ Versette™ Automated Liquid Handler
Support a wide range of applications with this compact handler, featuring user-friendly programming, reliable performance and a choice of 96- or 384-channel pipetting heads.
6145.00 € - 49445.00 €
Specifications
Channels 96, 384
Volume (Metric) 0.5 to 300 μL
View More Specs
Products 5
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
11684912
View Documents
Thermo Scientific™
650-01-BS
Versette Base Unit Only This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15118142
View Documents
Thermo Scientific™
650-02-NTC
96- and 384- channel pipetting module for use with 96- and 384-channel Versette pipetting heads This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11614922
View Documents
Thermo Scientific™
650-03-SPS
6-position stage for Versette, guarding included This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15453770
View Documents
Thermo Scientific™
650-MSIA
Versette system for MSIA application (includes base, pipetting module, 6-position stage, 96-channel pipetting head for volumes of 5-300μl and a serial dilute magazine 96/300μl) This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15443770
View Documents
Thermo Scientific™
650-INSTR
Versette instrument (includes base, pipetting module and a 6-position stage) This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Support a wide range of applications with the Thermo Scientific™ Versette™ Automated Liquid Handler, a compact handler featuring user-friendly programming, reliable performance and a choice of 96- or 384-channel pipetting heads. An ideal choice for the liquid handling needs of most laboratories, it supports 96/384 plate replication, plate stamping and serial dilution procedures.
Support a wide range of applications, including high-throughput Mass Spectrometric Immunoassay (MSIA), plate stamping, plate reformatting, serial dilution and many other applications.
96-and 384-channel pipetting heads:
  • Total volume range 0.5-300μL
  • All pipetting heads include RFID tags for self-identification
Consumables for optimized performance:
  • All pipetting heads utilize Thermo Scientific™ D.A.R.T.'S
Compact size:
  • Six position stage uses a unique dual-level design to minimize space requirements
  • Fits on standard bench
User-friendly programming options:
  • Simple pipetting procedures can be created using visual on-board graphical display
  • For more complex protocols and advanced editing, use Thermo Scientific™ ControlMate™ software included with purchase of the unit
  • Intuitive software; after minimal training, no specialist needed for protocol writing and revisions
  • Free WebEx ControlMate SW tutorial and applications support
Specifications
Specifications
96, 384
0.5 to 300 μL