missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Verso 1-Step RT-PCR Kit ReddyMix, with ThermoPrime Taq

Thermo Scientific Verso 1-Step RT-PCR Kit ReddyMix with ThermoPrime Taq is a complete set of RT-PCR reagents that combines everything needed for RT-PCR.

Brand:  Thermo Scientific™ AB1454LDB

Product Code. 11872113

  • 1452.00 € / 400 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific Verso 1-Step RT-PCR Kit ReddyMix with ThermoPrime Taq is a complete set of RT-PCR reagents that combines everything needed for RT-PCR. The Verso RT enzyme delivers high processivity and can be used at temperatures up to 57°C, delivering effective transcription through difficult RNA secondary structures. Verso RT also shows a better dynamic range than standard AMV or M-MuLV enzymes, with increases in template concentration giving rise to corresponding increases in product generated over a wider range of starting material concentrations.

An optional RT enhancer is provided with the kit and can be used during the RT step to remove any DNA contamination, eliminating the need for DNase I treatment.

This Verso 1-step RT-PCR system comes with the ReddyMix option, where the master mix contains a dye and precipitant that allow RT-PCR reactions to be loaded directly onto an electrophoresis gel. The red dye migrates between bromophenol blue and xylene cyanol at approximately 300 bp, depending on agarose concentration.

Features of the Verso 1-Step RT-PCR Kit ReddyMix with ThermoPrime Taq include:
• ReddyMix formulation—eliminates post-PCR steps required for electrophoresis
• Eliminates the need for DNase treatments—RT Enhancer (optional) prevents gDNA carry-over
• RT and PCR steps performed in single sealed tube. providing convenience and reduced contamination risk
• Verso RT enzyme is highly sensitive with a broad dynamic range—accurately detect RNA input from 1 pg to 1 μg
• Wide working temperature range (42–57°C) for success with GC-rich and other difficult templates
• RNase Inhibitor included in RT enzyme mix

Applications
• Gene expression analysis
• Viral/bacterial detection
• 1-step RT-PCR
• High throughput applications

Related products
Verso 1-Step RT-PCR Kit ReddyMix, with ThermoPrime Taq (Cat. No. AB1454LDA)
Verso 1-Step RT-PCR Kit, with Thermo-Start Taq (Hot Start) (Cat. No. AB1455A)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• Verso Enzyme Mix
• 2X Master Mix
• RT Enhancer

Store at –20°C.

Kit
ThermoPrime Taq DNA Polymerase
42°C to 57°C
Verso
Dry Ice
PCR Amplified cDNA
Premixed Components
Up to 11 kb
Gel Electrophoresis
1-step RT-PCR
Reverse Transcription
200 reactions
Yes
RT-PCR
200 Reactions
Reverse Transcription
RNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.