missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Verti-Gel™ Mini Tetrad PAGE system
Durable and leakproof system with capacity to run up to four gels.
631.00 € - 877.00 €
Specifications
Device Holds | 80 Samples, 20 Samples per Gel |
---|---|
Type | Verti-Gel Mini Tetrad PAGE System |
Material | Injection-molded |
No. of Gels | 4 |
Volume (Metric) Buffer | 250 to 1200 mL |
Product Code | Brand | Description | Includes | Thickness (Metric) Spacer | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Includes | Thickness (Metric) Spacer | Price | Quantity & Availability | |||
15126624
|
Fisherbrand™
FCVS10TETRAD1CBS |
With blotting module 100 x 100mm, 1mm spacer, 1mm comb | Blotting module 100 x 100mm, caster and external stand, four notched/plain plates with 1mm spacer, four plates, four 12-well 1mm comb, four cassette, eight pads | 1 mm spacer |
877.00 €
Each |
Estimated Shipment: 14-02-2025
Log in to see stock. |
|||
15146624
|
Fisherbrand™
FCVS10TETRAD1.5 |
With sliding clamps 100 x 100mm, 1.5mm bonded spacer, four 12-well 1.5mm combs | Sliding clamps 100 x 100mm,caster and external stand, 4x notched/plain plates with 1.5mm bonded spacers, four notched plates, four 12-well 1.5mm combs | 1.5 mm bonded spacer |
631.00 €
Each |
Estimated Shipment: 14-02-2025
Log in to see stock. |
|||
Description
- Injection-moulded construction
- COmpatible with 80 x 100mm and 100 x 100mm precast gels
- Low buffer volumes
- Interchangable modules for IEF/2D electrophoresis and electroblotting in universal tank
Specifications
80 Samples, 20 Samples per Gel | |
Injection-molded | |
250 to 1200 mL |
Verti-Gel Mini Tetrad PAGE System | |
4 | |
190 x 130 x 150 mm |