missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Vertical Gel Tank

Incorporates a sealing system which is compatible with all major types of 80mm x 100mm and 100mm x 100mm precast gel. Fisherbrand™ Vertical Gel Tank's ultra soft silicone seals and pressure bars which surround the glass plates guarantee leak proof gel casting.

286.00 € - 995.00 €

Specifications

For Use With (Equipment) 80 mm x 100 mm and 100 mm x 100 mm PreCast Gels
View More Specs

Products 5
Product Code Brand Weight (Metric) Includes Price Quantity & Availability  
Product Code Brand Weight (Metric) Includes Price Quantity & Availability  
11837623
View accessory products View Documents
Fisherbrand™
VS10BI
0.01 kg Blotting module Verti-gel mini with 4 cassettes and fibre pads
286.00 €
Each
In Stock
to see stock.
Edge
Add to basket
 
11893293
View accessory products View Documents
Fisherbrand™
FCVS10PRE
1.7 kg X1 Verti-Gel mini vertical system, no casting accessories
373.00 €
Each
Estimated Shipment: 23-01-2025
to see stock.
Add to basket
 
11883293
View accessory products View Documents
Fisherbrand™
FCVS10CBS
2.6 kg 1 Set Verti-Gel mini Complete vertical system for Electrophoresis and Blotting
995.00 €
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
 
11843293
View accessory products View Documents
Fisherbrand™
FCVS10DSYS
2.2 kg 1 Set Verti-Gel mini Complete vertical system for Electrophoresis
544.00 €
Each
Estimated Shipment: 06-02-2025
to see stock.
Add to basket
 
12623546
View Documents
Fisherbrand™
FVS20WAVESYS
0.01 kg 20x20cm plates, 2x 1mm 24 well combs,cooling coil,casting base
949.00 €
Each
Estimated Shipment: 04-03-2025
to see stock.
Add to basket
 
Description

Description

Incorporates a sealing system which is compatible with all major types of 80mm x 100mm and 100mm x 100mm precast gel. Gel casting and running utilise the same insert, no transfer of glass plates during gel casting is necessary. The insert contains sliding clamps allowing very rapid set up of both hand cast and precast gels. Ultra soft silicone seals and pressure bars which surround the glass plates guarantee leak proof gel casting. 2mm thick glass plates prevent breakage and have bonded spacers for convenience. Accessory electroblotting and tube gel modules are available which use the same outer tank and lid.
  • Injection molded construction, durable and leakproof
  • Compatible with all 80mm x 100mm and 100mm x 100mm precast gels
  • Low buffer volumes
  • Run up to four gels
  • Interchangable modules for IEF/2D electrophoresis and electroblotting in a universal tank For Mini SDS PAGE, native PAGE, gradient, second dimension and nucleic acid separations.
Specifications

Specifications

80 mm x 100 mm and 100 mm x 100 mm PreCast Gels
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.