Learn More
Contec™ VertiKlean™ MAX™ Hands-Free Mop Removal Tool
BIOPHARMAGREENER_CHOICE
Allows for hands free mop removal and disposal of all versions of Contec’s VertiKlean MAX and VertiKlean MAX Sealed Edge style mops.
Brand:  Contec™ 2639
158.00 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Product Code. 16283962
  • 222.00 € / Each
Estimated Shipment: 07-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • VertiKlean MAX Hands-Free Mop Removal Tool.
  • For Compact Bucket System.
Specifications
Specifications
Mop Head Removal Tool, Waste Bag Holder
Non-sterile
Reusable
VertiKlean MAX Hands-Free Mop Removal Tool
Stainless Steel
White
Not Autoclavable
Compact Bucket System
No