Learn More
Tocris Bioscience™ VH 032, thiol
Functionalized VHL ligand for PROTAC™; development
Brand:  Tocris Bioscience™ 7861/50
467.38 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 30000015
  • 592.00 € / 50mg
Explore more special offers
Description
Description
VH 032, thiol is a functionalized von-Hippel-Lindau protein ligand (VHL) for PROTAC® research and development; incorporates an E3 ligase ligand with thiol group for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
(2S,4R)-1-((R)-2-Acetamido-3-mercapto-3-methylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide
50 mg
C23H30N4O4S2
2098836-54-3
Degrader Building Blocks
0.95