Learn More
Tocris Bioscience™ VH 101, thiol
Thiol functionalized VHL ligand
Brand:  Tocris Bioscience™ 7884/50
638.43 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 30000031
  • 808.00 € / 50mg
Explore more special offers
Description
Description
VH 101, thiol is a functionalized von-Hippel-Lindau protein ligand (VHL) for PROTAC® research and development; incorporates an E3 ligase ligand with terminal thiol ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
(2S,4R)-1-[(2R)-2-[[(1-Fluorocyclopropyl)carbonyl]amino]-3-methyl-3-sulfanyl-butanoyl]-4-hydroxy-N-[[4-(4-methylthiazol-5-yl)phenyl]methyl]pyrrolidine-2-carboxamide
50 mg
C25H31FN4O4S2
2358775-81-0
Degrader Building Blocks
0.95