Learn More
Tocris Bioscience™ VH 298
High-affinity inhibitor of VHL
Brand:  Tocris Bioscience™ 6156/50
1021.14 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18770268
  • 1335.00 € / 50mg
Explore more special offers
Description
Description
VH 298 is a high-affinity inhibitor of E3 ubiquitin ligase VHL (Kd = 80-90 nM). Blocks interaction between VHL and HIF-α downstream of HIF-α hydroxylation, initiating hypoxic response. Results in time- and concentration-dependent accumulation of hydroxylated HIF-α, and upregulates mRNA and protein levels of HIF target genes, with similar transcriptional profile to hypoxia. Cell permeable.
Negative control cis VH 298 also available.
Specifications
Specifications
Inhibitor
C27H33N5O4S
Store at -20°C
50 mg
Ubiquitin Ligase (E3)
523.65g/mol
Ubiquitin Ligase (E3) Inhibitors