missing translation for 'onlineSavingsMsg'
Learn More

Block Technology VHP Decontamination Lock

Material airlock providing rapid H2O2 decontamination. Decontamination locks are most frequently used as a pass-through to transfer material between rooms with different classes of cleanliness (typically B and C). Loaded materials are decontaminated using hydrogen peroxide vapor.

Brand:  Block Technology VHPDL

Product Code. 17381689

  • POA / Each

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Integrated VHP generator
  • Decontamination pass-through cabin may be fitted onto the wall separating individual rooms
  • Lock complies with the leak tightness classes pursuant to ISO 10648-2
  • 'B' class cleanliness according to the EU GMP Standard
  • Electronic control system for automatic adjustment of basic operating modes managed by Siemens PLC
  • Color touchscreen controls
  • Mobile perforated shelves allow sterilization of the product from all sides
  • Process works at normal temperatures (at about 25 to 30°C) and normal pressures
  • Jacket material – stainless steel AISI 304
  • Pass-through chamber material – AISI 316L with a thickness of 3.00 mm, polished surface finish, Ra < 0.6 μm
  • Rounded corners R = 55 mm
  • Inlet and outlet H14 HEPA filtration
  • Possible validation of decontamination cycle
  • Possible data recording on a recording unit, printer or PC
  • Low operating costs
  • Decontamination a low-temperature
  • Quick interval of a decontamination cycle
  • Long operation life, minimal maintenance demands
  • Easy handling and operation

Operational Readiness - For operation of the equipment, the following must be ensured:

  • Pressurised air of pharmaceutical quality (dry, oil-free, 4 bars)
  • Electric power supply – 230 V, 3 kW
  • Connection to the exhaust branch for venting hydrogen peroxide vapors

Standards

  • Chamber closing door - It is made of PMMA with a thickness of 25 mm. The door is sealed using a pneumatic seal made of silicon. The closing of the door is monitored by a sensor; another sensor monitors the mechanism which prevents the door from being opened.
  • VHP distribution - Distribution pipe for optimal gassing inside the chamber
  • Perforated shelves - 4 pcs of standard perforated shelves for loaded material

Options

  • H2O2 concentration sensor – high – process sensor inside the chamber
  • H2O2 concentration sensor – low – safety sensor inside the chamber
  • H2O2 concentration sensor – low – safety sensor outside the chambre

Safety Features

  • Pressure fuse
Specifications

Specifications

Decontamination Lock
800 mm
1200 mm
Stainless Steel (316L)
2400 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers