Learn More
Proteintech Viability/Cytotoxicity Assay Kit for Animal Live & Dead Cells (Calcein AM, PI Method)
Viability/Cytotoxicity Assay Kit for Animal Live Dead Cells (Calcein AM, PI method) provides dual fluorescence staining for the detection of living and dead cells.
Brand:  Proteintech PF00007-150T
 View more versions of this product
Product Code. 17551176
  • 105.00 € / 150 tests
Explore more special offers
Description
Description
  • The two probes in the kit effect cell viability by measuring intracellular esterase activity and plasma membrane integrity.
  • This kit can be used for fluorescence microscopes, flow cytometers, microplate readers and other fluorescence detection systems.
  • This kit can be used for most eukaryotic mammalian samples, but not to fungi and yeast.
Specifications
Specifications
Flow Cytometry
617 nm, 517 nm
150 Tests