missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Vial, Cuvette and Test Tube Holders for Orion™ AQ7100 and AQ8100 Spectrophotometers

Quickly adapt your Orion™ AquaMate 7100 & 8100 Spectrophotometer to work with multiple sample vessels

Brand:  Thermo Scientific™ AQX1SQVH

 View more versions of this product

Product Code. 16469835

  • 25.93 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Easy to change & clean Orion™ AquaMate 7100 & 8100 Spectrophotometer sample holders are designed for maximum flexibility.

Easily adapt your Orion™ AquaMate 7100 & 8100 Spectrophotometer to work with multiple sample vessels

Magnetic retention makes these holders quick and easy to clean and change to adapt to your analytical needs.

  • AQX1RNDVH: Vial / Test Tube Holder for vials and test tube from 12 to 25 mm OD for Orion™ AquaMate 7100 & 8100 Spectrophotometers
  • AQX1SQVH: Standard 10 mm square single-cell holder for Orion™ AquaMate 7100 & 8100 Spectrophotometers
  • AQX1LWLVH: Long path rectangular cell holder for 20-100 mm pathlength cuvettes for Orion™ AquaMate 7100 & 8100 Spectrophotometers
TRUSTED_SUSTAINABILITY
Specifications

Specifications

10 mm square single-cell holder
Orion AquaMate 7100 & 8100 Spectrophotometers
1 square cell holder
Single Cell Sample Holder
10 mm cuvette (1)
Square
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers