Learn More
Veolia Sievers Certified TOC Vials
Use these certified TOC vials in critical pharmaceutical applications such as USP and EP Water testing as well as municipal water testing. Veolia Sievers Certified TOC Vials are certified to below 10 ppb for confidence in the quality of your TOC measurements.
Brand:  Veolia CHMI90606
Product Code. 13287930
  • 593.62 € / Pack of 72
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Sievers Certified (<10 ppb TOC) Vials
  • 72 x 40mL glass vials with caps and Teflon-lined septa, 72 per case
  • Ideal for strict regulatory compliance testing
  • Scrupulously cleaned in an ISO 9001 environment using a validated, automated process
  • Manufactured by the global leader in TOC measurement technology and TOC standards
  • Economical, with added savings on volume orders
  • Reliable, with guaranteed performance
  • Compatible with most TOC analyzer brands, including Sievers, Shimadzu, OI Analytical, Teledyne-Tekmar and Skalar
Specifications
Specifications
Clear
40 mL
72
Glass
Certified for use in critical applications including USP, EP, JP, and KP water testing
TOC Certified (<10 ppb)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Veolia Sievers Certified TOC Vials > Capacity: 40mL