Learn More
Heidolph™ Vibramax 100
Compact model with a load capacity of 2 kg.
Brand: Heidolph™ 544-21200-07
1111.50 EUR valid until 2025-12-31
Use promo code "24300" to get your promotional price.
This item is not returnable.
View return policy
Description
- Due to the 3 mm orbit and the speed of 150 to 1350 rpm, this circular vibrating shaker is ideal for suspending, for solubility studies and leaching studies.
- The optional tension roller attachment offers the option of using different container sizes. In combination with the non-slip rubber mat, the vessels are shaken safely even at high speeds. In addition, a timer with up to 120 minutes is integrated, which triggers an acoustic signal after expiration.
Specifications
230 V | |
UK Plug |