missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vici Valco Instruments™ Vici Valco Instruments Yellow PEEK Fitting
Click to view available options
Thread Size:
10-32
Unit Size:
Each
Specifications
Specifications
| Type | Fitting |
| Color | Yellow |
| For Use With (Application) | Can be Used in Virtually Any Type of 10-32 Thread HPLC Fitting |
| Packaging Quantity | 5 Pack |
| Thread Size | 10-32 |
| Pressure | 5000 psi |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction