missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Vilazodone hydrochloride
Low molecular weight, organic compound that may help regulate biological process.
63.00 € - 180.00 €
Specifications
Product Type | Agonist |
---|---|
Target Proteins | 5-HT1A |
CAS | 163521-08-2 |
Molecular Formula | C26H27N5O2.HCl |
Formula Weight | 477.99g/mol |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18785299
|
Tocris Bioscience™
6202/10 |
10 mg |
63.00 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18734868
|
Tocris Bioscience™
6202/50 |
50 mg |
180.00 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent 5-HT1A partial agonist and serotonin-selective reuptake inhibitor (SSRI) (IC50 values are 0.2 and 0.5 nM respectively). Antidepressant.Specifications
Agonist | |
163521-08-2 | |
477.99g/mol | |
5-HT1A Receptor Antagonists |
5-HT1A | |
C26H27N5O2.HCl | |
Store at -20°C |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title