missing translation for 'onlineSavingsMsg'
Learn More

Vinyl acetate, 99%, stab. with 8-12ppm hydroquinone

Product Code. 11463890
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
This item is not returnable. View return policy

Product Code. 11463890

Brand: Thermo Scientific Alfa Aesar A16247.AE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-05-4 | C4H6O2 | 86.09 g/mol

Vinyl acetate is used in the preparation of polyvinyl acetate emulsions and polyvinyl alcohol. It acts as a precursor to polyvinyl acetate. It serves as an ingredient in furniture-glue. It is also used to prepare copolymers with other monomers such as ethylene, acrylic acid. Further, it is used to make adhesives, paints and plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vinyl acetate is used in the preparation of polyvinyl acetate emulsions and polyvinyl alcohol. It acts as a precursor to polyvinyl acetate. It serves as an ingredient in furniture-glue. It is also used to prepare copolymers with other monomers such as ethylene, acrylic acid. Further, it is used to make adhesives, paints and plastics.

Solubility
Slightly miscible with water.

Notes
Light sensitive. Incompatible with acids, bases, oxidizing agents, peroxides, chlorosulfonic acid, ethylene imine, hydrochloric acid, oleum, nitric acid, sulfuric acid and 2-aminoethanol.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-05-4
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00008713
InChI Key XTXRWKRVRITETP-UHFFFAOYSA-N
Synonym vinyl acetate, acetic acid ethenyl ester, acetic acid vinyl ester, ethenyl ethanoate, 1-acetoxyethylene, vinyl ethanoate, acetoxyethene, vinylacetat, vinyl acetate monomer, vinyl a monomer
PubChem CID 7904
ChEBI CHEBI:46916
IUPAC Name ethenyl acetate
SMILES CC(=O)OC=C

Specifications

Melting Point -100°C
Density 0.932
Boiling Point 72°C to 73°C
Flash Point −8°C (18°F)
Odor Sweet
Linear Formula CH2=CHO2CCH3
Refractive Index 1.396
Quantity 100 mL
UN Number UN1301
Beilstein 1209327
Merck Index 14,9992
Solubility Information Slightly miscible with water.
Formula Weight 86.09
Percent Purity 99%
Chemical Name or Material Vinyl acetate, stabilized with 8 to 12ppm hydroquinone
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Vinyl acetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Vinyl acetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.