missing translation for 'onlineSavingsMsg'
Learn More

Vinyl bromide, 1M solution in THF, AcroSeal™

Product Code. p-4089502 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
This item is not returnable. View return policy

Product Code. 10339877

Brand: Thermo Scientific Acros 428771000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 593-60-2 | C2H3Br | 106.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Vinyl bromide
Name Note 1M solution in THF
CAS 593-60-2, 109-99-9
Density 0.9270g/mL
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C2H3Br
Linear Formula H2C=CHBr
MDL Number MFCD00000183
Quantity 100 mL
Specific Gravity 0.927
Synonym vinyl bromide, bromoethylene, ethene, bromo, monobromoethylene, vinylbromid, bromure de vinyle, vinylbromide, polyvinyl bromide, ethylene, bromo, polyvinylbromide
InChI Key INLLPKCGLOXCIV-UHFFFAOYSA-N
SMILES BrC=C
IUPAC Name bromoethene
Molecular Weight (g/mol) 106.95
PubChem CID 11641
ChEBI CHEBI:51311
Formula Weight 106.95
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Vinyl bromide, 1M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H350-May cause cancer.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H224-Extremely flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.