Learn More
3M™ Vinyl Tape 471F
Commonly used for marking hazards and aisles, color-coding, paint masking, decorative trim, splicing, and more. Made from vinyl backing with rubber adhesive, our versatile tape resists scrapes, wear, and weathering for long lasting use.
Brand:  3M™ 7000028853
 View more versions of this product
Product Code. 17287770
  • 1440.00 € / 24 rolls
Estimated Shipment: 27-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Maintains its color when exposed to abrasion, wear and many solvents
  • Unique stretch properties enable tape to remain stretched to conform to uneven surfaces without lifting and retracting
  • Ideal for use on curved and irregular surfaces
  • Easy to apply, excellent holding strength on many surfaces
  • Available in many distinctive colours (yellow, white, red, black, brown, green, orange, purple, blue and transparent)
  • Available in a wide variety of vivid colours and sizes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 mm x 33 m
32.9 m
5.1 cm
0.1 mm
0.14 mm
White